Assay Details
| Gene Symbol: GBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK002150.1 | - | 1 | 1646 | 92 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | guanylate binding protein 1 | GBP1 | -49.3kb | Sense | Chr. 1 - 89052304 - 89065360 on NCBI Build GRCh38 | |
| Downstream | guanylate binding protein 7 | GBP7 | GBP4L | 15.4kb | Sense | Chr. 1 - 89131751 - 89176040 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

GBP2
Genomic Map