Assay Details
| Gene Symbol: DNAJC21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK022694.1 | - | 1 | 545 | 121 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
BRX1, biogenesis of ribosomes |
BRIX1 |
BRIX, BXDC2 |
-26.5kb | Sense | Chr. 5 - 34915715 - 34925682 on NCBI Build GRCh38 |
| Downstream | alanine--glyoxylate aminotransferase 2 | AGXT2 |
AGT2, DAIBAT |
44.2kb | Anti-sense | Chr. 5 - 34998101 - 35048188 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000342382.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000342382.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000342382.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000342382.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000342382.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000342382.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000342382.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000342382.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000342382.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000342382.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000342382.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000342382.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000342382.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000342382.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000342382.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000342382.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000342382.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000382021.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000382021.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000382021.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000382021.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000382021.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000382021.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000382021.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000382021.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000382021.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000382021.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000382021.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000382021.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000382021.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000382021.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000382021.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000382021.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000382021.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000382021.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000382021.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000506762.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000506762.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000506762.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000509626.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000509626.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000509626.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000509626.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000512136.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000512136.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000512136.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000512136.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000514237.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000514237.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000514237.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000514237.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000514237.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000514237.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000514237.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000514237.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000514237.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | NM_001012339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | NM_001012339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | NM_001012339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | NM_001012339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | NM_001012339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | NM_001012339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | NM_001012339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | NM_001012339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | NM_001012339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | NM_001012339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | NM_001012339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | NM_001012339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | NM_001012339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | NM_001012339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | NM_001012339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | NM_001012339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | NM_001012339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | NM_001012339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | NM_194283.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | NM_194283.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | NM_194283.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | NM_194283.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | NM_194283.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | NM_194283.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | NM_194283.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | NM_194283.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | NM_194283.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | NM_194283.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | NM_194283.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | NM_194283.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | NM_194283.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | NM_194283.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | NM_194283.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | NM_194283.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | NM_194283.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | NM_194283.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | NM_194283.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000342382.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000342382.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000342382.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000342382.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000342382.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000342382.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000342382.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000342382.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000342382.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000342382.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000342382.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000342382.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000342382.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000342382.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000342382.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000342382.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000342382.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000382021.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000382021.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000382021.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000382021.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000382021.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000382021.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000382021.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000382021.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000382021.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000382021.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000382021.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000382021.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000382021.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000382021.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000382021.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000382021.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000382021.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000382021.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000382021.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000506762.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000506762.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000506762.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000509626.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000509626.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000509626.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000509626.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000512136.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000512136.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000512136.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000512136.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000514237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000514237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000514237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000514237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000514237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000514237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000514237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000514237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000514237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | NM_001012339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | NM_001012339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | NM_001012339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | NM_001012339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | NM_001012339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | NM_001012339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | NM_001012339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | NM_001012339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | NM_001012339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | NM_001012339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | NM_001012339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | NM_001012339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | NM_001012339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | NM_001012339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | NM_001012339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | NM_001012339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | NM_001012339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | NM_001012339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | NM_194283.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | NM_194283.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | NM_194283.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | NM_194283.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | NM_194283.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | NM_194283.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | NM_194283.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | NM_194283.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | NM_194283.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | NM_194283.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | NM_194283.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | NM_194283.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | NM_194283.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | NM_194283.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | NM_194283.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | NM_194283.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | NM_194283.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | NM_194283.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | NM_194283.3 | Refseq |

DNAJC21
Genomic Map