Assay Details
| Gene Symbol: SORT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK023648.1 | - | 1 | 1351 | 94 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | myosin binding protein H like | MYBPHL | -15.7kb | Sense | Chr. 1 - 109292365 - 109309394 on NCBI Build GRCh38 | |
| Downstream | proteasome subunit alpha 5 | PSMA5 |
PSC5, ZETA |
74.5kb | Sense | Chr. 1 - 109399031 - 109426486 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

SORT1
Genomic Map