Assay Details
| Gene Symbol: GATA2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK024653.1 | - | 1 | 1252 | 91 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | GATA binding protein 2 | GATA2 |
DCML, IMD21, MONOMAC, NFE1B |
-5.4kb | Anti-sense | Chr. 3 - 128479422 - 128493187 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC90246 | LOC90246 | 7.5kb | Sense | Chr. 3 - 128507835 - 128510595 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

GATA2-AS1
Genomic Map