Assay Details
| Gene Symbol: OSBPL10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK024945.1 | - | 1 | 1046 | 86 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | OSBPL10 antisense RNA 1 | OSBPL10-AS1 | -181.7kb | Anti-sense | Chr. 3 - 31704213 - 31721586 on NCBI Build GRCh38 | |
| Downstream | zinc finger protein 860 | ZNF860 | 76.8kb | Anti-sense | Chr. 3 - 31981774 - 32005112 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

OSBPL10
Genomic Map