Assay Details
| Gene Symbol: ROPN1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK026656.1 | - | 1 | 1350 | 96 | |
| Gene Symbol: LOC105369214 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011514190.2 | XP_011512492.1 | 1 | 1656 | 96 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ROPN1L antisense RNA 1 | ROPN1L-AS1 | -21.3kb | Anti-sense | Chr. 5 - 10440735 - 10441851 on NCBI Build GRCh38 | |
| Downstream | microRNA 6131 | MIR6131 | hsa-mir-6131 | 13.0kb | Sense | Chr. 5 - 10478037 - 10478145 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

ROPN1L
Genomic Map