Assay Details
| Gene Symbol: TRIM9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011536389.2 | XP_011534691.1 | 13 | 3899 | 102 | |
| GenBank mRNA | AK054781.1 | - | 1 | 1855 | 102 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC400212 | LOC400212 | -16.8kb | Sense | Chr. 14 - 50946296 - 50962321 on NCBI Build GRCh38 | |
| Downstream | thioredoxin related transmembrane protein 1 | TMX1 |
PDIA11, TMX, TXNDC, TXNDC1 |
259.1kb | Anti-sense | Chr. 14 - 51240168 - 51257655 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

TRIM9
Genomic Map