Assay Details
| Gene Symbol: NEURL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285486.1 | NP_001272415.1 | 2 - 3 | 302 | 91 | |
| GenBank mRNA | AL389981.1 | - | 2 - 3 | 262 | 91 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | non-SMC condensin I complex subunit H | NCAPH |
BRRN1, CAP-H |
-123.3kb | Anti-sense | Chr. 2 - 96335741 - 96377095 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC105373496 | LOC105373496 | 23.2kb | Sense | Chr. 2 - 96527940 - 96532306 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the NM_001285486.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | ENST00000310865.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | ENST00000310865.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | ENST00000310865.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | ENST00000310865.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | ENST00000310865.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | ENST00000310865.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | ENST00000310865.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | ENST00000310865.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | ENST00000310865.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | ENST00000310865.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | ENST00000310865.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | ENST00000435380.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | ENST00000435380.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | ENST00000435380.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | ENST00000435380.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | ENST00000435380.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | ENST00000435380.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | ENST00000435380.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | ENST00000444588.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | ENST00000444588.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | ENST00000444588.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | ENST00000444588.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | ENST00000444588.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | ENST00000451794.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | ENST00000451794.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | ENST00000451794.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | ENST00000451794.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | ENST00000451794.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | ENST00000451794.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | ENST00000451794.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | ENST00000451794.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | NM_001285485.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | NM_001285485.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | NM_001285485.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | NM_001285485.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | NM_001285485.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | NM_001285485.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | NM_001285485.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | NM_001285485.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | NM_001285485.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | NM_001285486.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | NM_001285486.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | NM_001285486.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | NM_001285486.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | NM_001285486.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | NM_001285486.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | NM_001285486.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | NM_001285486.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | NM_001285486.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | NM_001285486.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | NM_001285486.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | NM_001285486.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | NR_104346.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | NR_104346.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | NR_104346.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | NR_104346.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | NR_104346.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | NR_104346.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | NR_104346.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200198954.hg.1 | ENST00000310865.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200198954.hg.1 | NM_001285486.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200198954.hg.1 | ENST00000310865.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200198954.hg.1 | NM_001285486.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200198954.hg.1 | TC0200013524.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | ENST00000310865.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | ENST00000310865.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | ENST00000310865.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | ENST00000310865.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | ENST00000310865.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | ENST00000310865.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | ENST00000310865.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | ENST00000310865.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | ENST00000310865.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | ENST00000310865.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | ENST00000310865.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | ENST00000435380.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | ENST00000435380.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | ENST00000435380.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | ENST00000435380.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | ENST00000435380.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | ENST00000435380.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | ENST00000435380.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | ENST00000444588.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | ENST00000444588.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | ENST00000444588.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | ENST00000444588.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | ENST00000444588.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | ENST00000451794.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | ENST00000451794.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | ENST00000451794.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | ENST00000451794.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | ENST00000451794.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | ENST00000451794.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | ENST00000451794.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | ENST00000451794.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | NM_001285485.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | NM_001285485.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | NM_001285485.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | NM_001285485.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | NM_001285485.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | NM_001285485.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | NM_001285485.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | NM_001285485.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | NM_001285485.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | NM_001285486.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | NM_001285486.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | NM_001285486.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | NM_001285486.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | NM_001285486.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | NM_001285486.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | NM_001285486.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | NM_001285486.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | NM_001285486.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | NM_001285486.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | NM_001285486.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | NM_001285486.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | NR_104346.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | NR_104346.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | NR_104346.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | NR_104346.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | NR_104346.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | NR_104346.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | NR_104346.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 173654 | - | - |

NEURL3
Genomic Map