Assay Details
| Gene Symbol: LINC00032 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_026679.1 | 1 - 2 | 468 | 69 | ||
| GenBank mRNA | AF418573.2 | - | 1 - 2 | 468 | 69 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09000986.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0900009774.hg.1 | ENST00000425633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009774.hg.1 | ENST00000425633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009774.hg.1 | ENST00000425633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009774.hg.1 | ENST00000425633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009774.hg.1 | ENST00000425633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009774.hg.1 | ENST00000425633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009774.hg.1 | ENST00000425633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009774.hg.1 | ENST00000425633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009774.hg.1 | ENST00000425633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009774.hg.1 | ENST00000425633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009774.hg.1 | ENST00000425633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009774.hg.1 | ENST00000425633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009774.hg.1 | ENST00000425633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009774.hg.1 | ENST00000425633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009774.hg.1 | ENST00000425633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009774.hg.1 | ENST00000425633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009774.hg.1 | ENST00000425633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009774.hg.1 | ENST00000425633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009774.hg.1 | NR_026679.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009774.hg.1 | NR_026679.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009774.hg.1 | NR_026679.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009774.hg.1 | NR_026679.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009774.hg.1 | NR_026679.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009774.hg.1 | NR_026679.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009774.hg.1 | NR_026679.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009774.hg.1 | NR_026679.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009774.hg.1 | NR_026679.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009774.hg.1 | NR_026679.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009774.hg.1 | NR_026679.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009774.hg.1 | NR_026679.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009774.hg.1 | NR_026679.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009774.hg.1 | NR_026679.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009774.hg.1 | NR_026679.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009774.hg.1 | NR_026679.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009774.hg.1 | NR_026679.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009774.hg.1 | NR_026679.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09012965.hg.1 | TC09000986.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09012965.hg.1 | ENST00000425633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09012965.hg.1 | NR_026679.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09012966.hg.1 | TC09000986.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09012966.hg.1 | ENST00000425633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09012966.hg.1 | NR_026679.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900166884.hg.1 | ENST00000425633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900166884.hg.1 | NR_026679.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900166884.hg.1 | ENST00000425633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900166884.hg.1 | NR_026679.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900166884.hg.1 | TC0900009774.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900166883.hg.1 | ENST00000425633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900166883.hg.1 | NR_026679.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900166883.hg.1 | ENST00000425633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900166883.hg.1 | NR_026679.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900166883.hg.1 | TC0900009774.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900009774.hg.1 | ENST00000425633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009774.hg.1 | ENST00000425633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009774.hg.1 | ENST00000425633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009774.hg.1 | ENST00000425633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009774.hg.1 | ENST00000425633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009774.hg.1 | ENST00000425633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009774.hg.1 | ENST00000425633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009774.hg.1 | ENST00000425633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009774.hg.1 | ENST00000425633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009774.hg.1 | ENST00000425633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009774.hg.1 | ENST00000425633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009774.hg.1 | ENST00000425633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009774.hg.1 | ENST00000425633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009774.hg.1 | ENST00000425633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009774.hg.1 | ENST00000425633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009774.hg.1 | ENST00000425633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009774.hg.1 | ENST00000425633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009774.hg.1 | ENST00000425633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009774.hg.1 | NR_026679.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009774.hg.1 | NR_026679.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009774.hg.1 | NR_026679.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009774.hg.1 | NR_026679.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009774.hg.1 | NR_026679.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009774.hg.1 | NR_026679.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009774.hg.1 | NR_026679.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009774.hg.1 | NR_026679.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009774.hg.1 | NR_026679.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009774.hg.1 | NR_026679.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009774.hg.1 | NR_026679.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009774.hg.1 | NR_026679.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009774.hg.1 | NR_026679.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009774.hg.1 | NR_026679.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009774.hg.1 | NR_026679.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009774.hg.1 | NR_026679.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009774.hg.1 | NR_026679.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009774.hg.1 | NR_026679.1 | Refseq |

LINC00032
Genomic Map