Assay Details
| Gene Symbol: DIO3OS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF469205.1 | - | 1 - 2 | 940 | 80 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC100507277 | LOC100507277 | -111.2kb | Anti-sense | Chr. 14 - 101442076 - 101444315 on NCBI Build GRCh38 | |
| Downstream | microRNA 1247 | MIR1247 |
MIRN1247, hsa-mir-1247, mir-1247 |
2.9kb | Sense | Chr. 14 - 101560287 - 101560422 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

DIO3OS
Genomic Map