Assay Details
| Gene Symbol: GPR12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005288.3 | NP_005279.1 | 2 | 3491 | 74 | |
| XM_005266360.3 | XP_005266417.1 | 2 | 3088 | 74 | ||
| GenBank mRNA | AK055229.1 | - | 1 | 2147 | 74 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | WAS protein family member 3 | WASF3 |
Brush-1, SCAR3, WAVE3 |
-67.4kb | Anti-sense | Chr. 13 - 26557703 - 26688945 on NCBI Build GRCh38 |
| Downstream | ubiquitin specific peptidase 12 | USP12 |
UBH1, USP12L1 |
307.4kb | Sense | Chr. 13 - 27066150 - 27171896 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

GPR12
Genomic Map