Assay Details
| Gene Symbol: NONO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK055406.1 | - | 1 | 648 | 98 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger MYM-type containing 3 | ZMYM3 |
DXS6673E, MYM, XFIM, ZNF198L2, ZNF261 |
-43.6kb | Anti-sense | Chr. X - 71239624 - 71255261 on NCBI Build GRCh38 |
| Downstream | integrin subunit beta 1 binding protein 2 | ITGB1BP2 |
CHORDC3, ITGB1BP, MELUSIN, MSTP015 |
582.0 | Sense | Chr. X - 71301275 - 71305371 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

NONO
Genomic Map