Assay Details
| Gene Symbol: LINC00895 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK057431.1 | - | 1 | 1248 | 96 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | claudin 5 | CLDN5 |
AWAL, BEC1, CPETRL1, TMVCF |
-39.9kb | Sense | Chr. 22 - 19523024 - 19527545 on NCBI Build GRCh38 |
| Downstream | septin 5 | SEPT5 |
CDCREL, CDCREL-1, CDCREL1, H5, HCDCREL-1, PNUTL1 |
147.6kb | Anti-sense | Chr. 22 - 19714464 - 19723322 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LINC00895
Genomic Map