Assay Details
Gene Symbol: M6PR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK057556.1 | - | 1 | 1945 | 93 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | polyhomeotic homolog 1 | PHC1 |
EDR1, HPH1, MCPH11, RAE28 |
-6.4kb | Anti-sense | Chr. 12 - 8914664 - 8941467 on NCBI Build GRCh38 |
Downstream | killer cell lectin like receptor G1 | KLRG1 |
2F1, CLEC15A, MAFA, MAFA-2F1, MAFA-L, MAFA-LIKE |
39.7kb | Anti-sense | Chr. 12 - 8989079 - 9255692 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000000412.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000000412.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000000412.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000000412.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000000412.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000000412.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000000412.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000000412.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000000412.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000000412.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000000412.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000000412.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000000412.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000000412.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000000412.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000000412.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000000412.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000000412.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000000412.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000000412.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000536844.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000536844.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000536844.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000536844.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000536844.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000536844.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000536844.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000536844.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000536844.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000536844.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000536844.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000536844.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000536844.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000536844.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000536844.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000537621.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000537621.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000537621.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000537621.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000537621.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000537621.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000539143.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000539143.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000539143.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000539143.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000539143.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000540837.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000540837.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000540837.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000540837.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000540837.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000540837.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000541507.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000541507.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000541507.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000541507.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000541507.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000541507.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000541507.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000541507.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000541507.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000541507.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000541507.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000541507.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000541507.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000541507.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000541507.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000543159.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000543159.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000543159.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000543159.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000543159.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000543258.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000543258.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000543258.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000543258.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000543258.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000543258.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000543258.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000543704.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000543704.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000543704.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000543704.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000543704.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000543704.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000543704.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000543834.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000543834.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000543834.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000543834.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000543845.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000543845.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000543845.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000543845.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000543845.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000543845.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000543845.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000544193.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000544193.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000544193.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000544193.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000544245.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000544245.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000544245.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000544245.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000544245.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000544245.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000544245.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000544245.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | NM_001207024.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | NM_001207024.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | NM_001207024.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | NM_001207024.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | NM_001207024.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | NM_001207024.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | NM_001207024.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | NM_001207024.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | NM_001207024.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | NM_001207024.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | NM_001207024.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | NM_001207024.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | NM_001207024.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | NM_001207024.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | NM_001207024.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | NM_002355.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | NM_002355.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | NM_002355.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | NM_002355.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | NM_002355.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | NM_002355.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | NM_002355.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | NM_002355.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | NM_002355.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | NM_002355.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | NM_002355.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | NM_002355.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | NM_002355.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | NM_002355.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | NM_002355.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | NM_002355.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | NM_002355.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | NM_002355.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | NM_002355.3 | Refseq |
