Assay Details
| Gene Symbol: LINC00221 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK058096.1 | - | 3 - 4 | 1040 | 84 | |
More Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09537243 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30005685 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09537224 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30005675 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16745760 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09537221 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09537225 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30005701 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09537216 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16745761 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30005711 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09537229 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30005684 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30005671 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09537237 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16745755 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30005683 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09537222 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09537228 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09537233 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09537210 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09537213 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30005712 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16745759 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30005708 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16745758 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30005692 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30005706 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30005709 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16745744 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16745753 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09537219 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30005681 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09537230 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 82174 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 381932 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 165006 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 905679 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 743741 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 346918 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 180794 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 332829 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 80837 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 295801 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 120535 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 330882 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 138998 | AL117638.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 78264 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 136851 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 258749 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 234813 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_32_P162192 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3464438 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 33387 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 18773 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 24273 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P02991750 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_32_P162192 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3464438 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_32_P162192 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30005633 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30005635 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16745717 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30005649 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16745726 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30005625 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09537202 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09537203 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30005623 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16745717 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30005633 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30005649 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30005635 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09537203 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16745726 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09537202 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30005623 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30005625 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09537260 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09537243 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16745746 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16745733 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16745755 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16745748 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09537212 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16745753 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09537210 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09537264 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09537273 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09537237 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09537239 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09537232 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16745754 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09537268 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09537219 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09537230 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09537255 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09537270 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09537257 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09537247 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09537252 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16745760 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16745744 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09537217 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09537249 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09537224 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09537214 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09537222 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09537228 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09537263 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09537266 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09537226 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09537203 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09537204 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P40323672 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P20176541 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P02991750 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 7977449 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7977448 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7977447 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 1564601_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009320.hg.1 | ENST00000557882.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009320.hg.1 | ENST00000557882.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009320.hg.1 | ENST00000557882.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009320.hg.1 | ENST00000557882.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009320.hg.1 | ENST00000557882.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009320.hg.1 | ENST00000557882.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009320.hg.1 | ENST00000557882.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009320.hg.1 | ENST00000557882.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009320.hg.1 | ENST00000559799.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009320.hg.1 | ENST00000559799.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009320.hg.1 | ENST00000559799.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009320.hg.1 | ENST00000559799.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009320.hg.1 | ENST00000559799.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009320.hg.1 | ENST00000559799.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009320.hg.1 | ENST00000559799.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009320.hg.1 | ENST00000559799.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009320.hg.1 | ENST00000559799.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009320.hg.1 | ENST00000559799.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009320.hg.1 | ENST00000560180.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009320.hg.1 | ENST00000560180.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009320.hg.1 | ENST00000560180.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009320.hg.1 | ENST00000560180.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009320.hg.1 | ENST00000560180.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008519.hg.1 | ENST00000334298.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008519.hg.1 | ENST00000334298.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008519.hg.1 | ENST00000334298.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008519.hg.1 | ENST00000433371.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008519.hg.1 | ENST00000433371.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008519.hg.1 | ENST00000433371.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008519.hg.1 | ENST00000433371.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008519.hg.1 | ENST00000449670.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008519.hg.1 | ENST00000449670.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008519.hg.1 | ENST00000449670.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008519.hg.1 | ENST00000603633.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008519.hg.1 | ENST00000603633.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008519.hg.1 | ENST00000603633.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008519.hg.1 | ENST00000619530.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008519.hg.1 | ENST00000619530.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008519.hg.1 | ENST00000619530.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008519.hg.1 | ENST00000619530.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008519.hg.1 | ENST00000620337.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008519.hg.1 | ENST00000620337.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008519.hg.1 | ENST00000620337.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009320.hg.1 | ENST00000557882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009320.hg.1 | ENST00000557882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009320.hg.1 | ENST00000557882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009320.hg.1 | ENST00000557882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009320.hg.1 | ENST00000557882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009320.hg.1 | ENST00000557882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009320.hg.1 | ENST00000557882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009320.hg.1 | ENST00000557882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009320.hg.1 | ENST00000559799.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009320.hg.1 | ENST00000559799.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009320.hg.1 | ENST00000559799.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009320.hg.1 | ENST00000559799.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009320.hg.1 | ENST00000559799.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009320.hg.1 | ENST00000559799.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009320.hg.1 | ENST00000559799.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009320.hg.1 | ENST00000559799.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009320.hg.1 | ENST00000559799.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009320.hg.1 | ENST00000559799.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009320.hg.1 | ENST00000560180.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009320.hg.1 | ENST00000560180.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009320.hg.1 | ENST00000560180.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009320.hg.1 | ENST00000560180.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009320.hg.1 | ENST00000560180.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008519.hg.1 | ENST00000334298.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008519.hg.1 | ENST00000334298.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008519.hg.1 | ENST00000334298.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008519.hg.1 | ENST00000433371.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008519.hg.1 | ENST00000433371.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008519.hg.1 | ENST00000433371.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008519.hg.1 | ENST00000433371.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008519.hg.1 | ENST00000449670.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008519.hg.1 | ENST00000449670.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008519.hg.1 | ENST00000449670.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008519.hg.1 | ENST00000603633.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008519.hg.1 | ENST00000603633.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008519.hg.1 | ENST00000603633.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008519.hg.1 | ENST00000619530.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008519.hg.1 | ENST00000619530.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008519.hg.1 | ENST00000619530.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008519.hg.1 | ENST00000619530.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008519.hg.1 | ENST00000620337.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008519.hg.1 | ENST00000620337.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008519.hg.1 | ENST00000620337.4 | Ensembl |
| Human Genome U133 Plus 2.0 Array | 1564601_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1564601_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564601_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564601_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564601_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564601_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564601_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564601_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564601_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564601_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564601_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564601_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564601_at1 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A051879 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A051879 | - | - |

LINC00221
Genomic Map