Assay Details
| Gene Symbol: C9orf131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK090398.1 | - | 1 | 1847 | 86 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | BF903811.1 | - | Chr. - - [-] on Build | 86 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | DnaJ heat shock protein family (Hsp40) member B5 | DNAJB5 | Hsc40 | -40.2kb | Anti-sense | Chr. 9 - 34989369 - 34998433 on NCBI Build GRCh38 |
| Downstream | valosin containing protein | VCP |
ALS14, CMT2Y, HEL-220, HEL-S-70, IBMPFD, IBMPFD1, TERA, p97 |
11.7kb | Sense | Chr. 9 - 35056068 - 35072742 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0900009926.hg.1 | ENST00000624351.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | ENST00000312292.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | ENST00000312292.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | ENST00000312292.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | ENST00000312292.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | ENST00000312292.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | ENST00000312292.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | ENST00000312292.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | ENST00000312292.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | ENST00000312292.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | ENST00000312292.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | ENST00000354479.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | ENST00000354479.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | ENST00000354479.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | ENST00000354479.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | ENST00000354479.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | ENST00000354479.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | ENST00000354479.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | ENST00000378745.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | ENST00000378745.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | ENST00000378745.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | ENST00000378745.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | ENST00000378745.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | ENST00000378745.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | ENST00000416537.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | ENST00000416537.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | ENST00000416537.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | ENST00000416537.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | ENST00000421362.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | ENST00000421362.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | ENST00000421362.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | ENST00000421362.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | ENST00000421362.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | ENST00000421362.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | ENST00000421362.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | ENST00000421362.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | ENST00000534880.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | ENST00000534880.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | ENST00000534880.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | ENST00000537671.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | ENST00000537671.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | ENST00000537671.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | NM_001040410.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | NM_001040410.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | NM_001040410.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | NM_001040410.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | NM_001040410.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | NM_001040410.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | NM_001040410.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | NM_001040410.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | NM_001040411.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | NM_001040411.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | NM_001040411.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | NM_001040411.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | NM_001040411.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | NM_001040411.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | NM_001040411.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | NM_001040412.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | NM_001040412.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | NM_001040412.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | NM_001040412.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | NM_001040412.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | NM_001040412.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | NM_001040412.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | NM_001040412.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | NM_001287391.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | NM_001287391.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | NM_001287391.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | NM_001287391.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | NM_001287391.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | NM_001287391.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | NM_001287391.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | NM_203299.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | NM_203299.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | NM_203299.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | NM_203299.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | NM_203299.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | NM_203299.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | NM_203299.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | NM_203299.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | NM_203299.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007069.hg.1 | NM_203299.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | ENST00000312292.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | ENST00000312292.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | ENST00000312292.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | ENST00000312292.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | ENST00000312292.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | ENST00000312292.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | ENST00000312292.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | ENST00000312292.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | ENST00000312292.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | ENST00000312292.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | ENST00000354479.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | ENST00000354479.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | ENST00000354479.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | ENST00000354479.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | ENST00000354479.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | ENST00000354479.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | ENST00000354479.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | ENST00000378745.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | ENST00000378745.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | ENST00000378745.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | ENST00000378745.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | ENST00000378745.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | ENST00000378745.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | ENST00000416537.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | ENST00000416537.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | ENST00000416537.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | ENST00000416537.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | ENST00000421362.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | ENST00000421362.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | ENST00000421362.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | ENST00000421362.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | ENST00000421362.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | ENST00000421362.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | ENST00000421362.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | ENST00000421362.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | ENST00000534880.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | ENST00000534880.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | ENST00000534880.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | ENST00000537671.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | ENST00000537671.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | ENST00000537671.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | NM_001040410.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | NM_001040410.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | NM_001040410.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | NM_001040410.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | NM_001040410.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | NM_001040410.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | NM_001040410.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | NM_001040410.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | NM_001040411.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | NM_001040411.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | NM_001040411.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | NM_001040411.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | NM_001040411.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | NM_001040411.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | NM_001040411.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | NM_001040412.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | NM_001040412.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | NM_001040412.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | NM_001040412.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | NM_001040412.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | NM_001040412.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | NM_001040412.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | NM_001040412.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | NM_001287391.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | NM_001287391.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | NM_001287391.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | NM_001287391.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | NM_001287391.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | NM_001287391.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | NM_001287391.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | NM_203299.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | NM_203299.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | NM_203299.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | NM_203299.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | NM_203299.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | NM_203299.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | NM_203299.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | NM_203299.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | NM_203299.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007069.hg.1 | NM_203299.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009926.hg.1 | ENST00000624351.1 | Ensembl |

C9orf131
Genomic Map