Assay Details
| Gene Symbol: PAPPA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK092441.1 | - | 1 | 1850 | 79 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 474 | LINC00474 |
C9orf27, EST-YD1 |
-422.3kb | Anti-sense | Chr. 9 - 115888265 - 115925098 on NCBI Build GRCh38 |
| Downstream | PAPPA antisense RNA 1 | PAPPA-AS1 |
DIPAS, NCRNA00156, PAPPA-AS, PAPPAAS, PAPPAS, TAF2C2, TAF4B, TAFII105 |
48.5kb | Anti-sense | Chr. 9 - 116398157 - 116400606 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

PAPPA
Genomic Map