Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P27554219 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16318411 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16318398 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16318399 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P07188526 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16318404 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P07188538 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P07188539 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P27554213 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16318421 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16318413 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P07188525 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16318364 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16318403 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P27554199 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P07188527 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16318397 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P07188533 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16318370 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P27554241 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P07188536 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16318387 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16318359 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P07188531 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16318410 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P27554240 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P07188530 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16318412 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16318406 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P07188535 |
- |
- |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
190420 |
BC132831.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
190420 |
BC132833.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
190420 |
BX647582.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
190420 |
NM_001012418.3 |
refseq_mrna_human_NM |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
151200 |
AL117627.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
151200 |
BC029927.1 |
genbank_mrna_human |
Agilent Whole Human Genome Microarray 4x44K v2 |
A_33_P3248231 |
- |
- |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
25128 |
- |
- |
SurePrint G3 Human GE Microarray G4851A |
A_33_P3248231 |
- |
- |