Assay Details
| Gene Symbol: IL7R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002185.3 | NP_002176.2 | 8 | 3559 | 105 | |
| NR_120485.1 | 6 | 3296 | 105 | |||
| XM_005248299.3 | XP_005248356.1 | 7 | 3450 | 105 | ||
| GenBank mRNA | AL713738.1 | - | 1 | 247 | 105 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | sperm flagellar 2 | SPEF2 |
CT122, KPL2 |
-63.7kb | Sense | Chr. 5 - 35617773 - 35814611 on NCBI Build GRCh38 |
| Downstream | calcyphosine like | CAPSL | 24.7kb | Anti-sense | Chr. 5 - 35904295 - 35938779 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

IL7R
Genomic Map