Assay Details
| Gene Symbol: AGBL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AL832281.1 | - | 1 | 1842 | 103 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | caldesmon 1 | CALD1 |
CDM, H-CAD, HCAD, L-CAD, LCAD, NAG22 |
-15.8kb | Sense | Chr. 7 - 134779413 - 134970729 on NCBI Build GRCh38 |
| Downstream | transmembrane protein 140 | TMEM140 | 158.3kb | Sense | Chr. 7 - 135148014 - 135166215 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

AGBL3
Genomic Map