Assay Details
| Gene Symbol: ATP2C1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AL833453.1 | - | 1 | 2551 | 86 | |
| Gene Symbol: ASTE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017006268.1 | XP_016861757.1 | 6 | 86 | ||
| XM_017006269.1 | XP_016861758.1 | 6 | 86 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | phosphoinositide-3-kinase regulatory subunit 4 | PIK3R4 |
VPS15, p150 |
-266.1kb | Anti-sense | Chr. 3 - 130678934 - 130746852 on NCBI Build GRCh38 |
| Downstream | NIMA related kinase 11 | NEK11 | 10.2kb | Sense | Chr. 3 - 131026850 - 131350465 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

ATP2C1
Genomic Map