Assay Details
| Gene Symbol: SFTA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY102071.1 | - | 1 - 2 | 52 | 65 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | NKX2-1 antisense RNA 1 | NKX2-1-AS1 | Anti-sense | Chr. 14 - 36519278 - 36523016 on NCBI Build GRCh38 | ||
| Downstream | NK2 homeobox 8 | NKX2-8 |
NKX2.8, NKX2H, Nkx2-9 |
60.5kb | Sense | Chr. 14 - 36580011 - 36582614 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

SFTA3
Genomic Map