Assay Details
| Gene Symbol: TEF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB051442.1 | - | 1 | 2357 | 92 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC100996598 | LOC100996598 | Sense | Chr. 22 - 41369082 - 41370396 on NCBI Build GRCh38 | ||
| Downstream |
transducer of ERBB2, 2 |
TOB2 |
APRO5, TOB4, TOBL, TROB2 |
59.8kb | Anti-sense | Chr. 22 - 41433488 - 41448230 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

TEF
Genomic Map