Assay Details
| Gene Symbol: KCNE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF086086.1 | - | 2 | 555 | 101 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 310 | LINC00310 |
C21orf82, NCRNA00310 |
-176.8kb | Sense | Chr. 21 - 34180677 - 34189919 on NCBI Build GRCh38 |
| Downstream | small integral membrane protein 11A | SMIM11A |
C21orf51, FAM165B, SMIM11 |
8.3kb | Sense | Chr. 21 - 34375450 - 34402777 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

KCNE2
Genomic Map