Assay Details
| Gene Symbol: RASGRF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF370414.1 | - | 1 | 1255 | 105 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cathepsin H | CTSH |
ACC-4, ACC-5, ACC4, ACC5, CPSB |
-33.1kb | Anti-sense | Chr. 15 - 78921750 - 78945098 on NCBI Build GRCh38 |
| Downstream | ANKRD34C antisense RNA 1 | ANKRD34C-AS1 | 211.5kb | Anti-sense | Chr. 15 - 79191707 - 79283945 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

RASGRF1
Genomic Map