Assay Details
| Gene Symbol: DCAF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK022145.1 | - | 2148 | 83 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | potassium voltage-gated channel subfamily H member 6 | KCNH6 |
ERG-2, ERG2, HERG2, Kv11.2, hERG-2 |
-36.5kb | Anti-sense | Chr. 17 - 63523334 - 63548977 on NCBI Build GRCh38 |
| Downstream | translational activator of cytochrome c oxidase I | TACO1 | CCDC44 | 15.0kb | Anti-sense | Chr. 17 - 63600872 - 63608365 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

DCAF7
Genomic Map