Assay Details
| Gene Symbol: MCF2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK057899.1 | - | 1 | 645 | 111 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | MCF2L antisense RNA 1 | MCF2L-AS1 | -123.6kb | Anti-sense | Chr. 13 - 112967484 - 112968638 on NCBI Build GRCh38 | |
| Downstream | coagulation factor VII | F7 | SPCA | 12.7kb | Sense | Chr. 13 - 113105773 - 113120681 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

MCF2L
Genomic Map