| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Gene 1.0 ST Array |
8104073 |
- |
- |
| Affymetrix Human Gene 1.1 ST Array |
8104072 |
- |
- |
| Affymetrix HT HG-U133+ PM Array Plate |
229959_PM_at |
- |
- |
| Affymetrix HT HG-U133+ PM Array Plate |
1559441_PM_s_at |
- |
- |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012663.hg.1 |
ENST00000623820.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012663.hg.1 |
NR_046264.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400186842.hg.1 |
ENST00000623820.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186842.hg.1 |
NR_046264.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400186842.hg.1 |
ENST00000623820.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400186842.hg.1 |
NR_046264.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400186842.hg.1 |
TC0400012663.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0400012663.hg.1 |
ENST00000623820.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012663.hg.1 |
NR_046264.1 |
Refseq |
| Human Genome U133 Plus 2.0 Array |
1559441_s_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
1559441_PM_s_at |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1559441_s_at7 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1559441_s_at10 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1559441_s_at3 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1559441_s_at4 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1559441_s_at6 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1559441_s_at11 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1559441_s_at2 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1559441_s_at8 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1559441_s_at5 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1559441_s_at1 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1559441_s_at9 |
- |
- |
| Human Genome U95 Set |
78984_PM_at |
- |
- |
| Human Genome U95 Set |
78984_at |
- |
- |