Assay Details
| Gene Symbol: LOC100130027 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK127225.1 | - | 5 | 2451 | 108 | |
| Gene Symbol: ZNF517 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011517014.2 | XP_011515316.1 | 5 | 108 | ||
| XM_011517015.2 | XP_011515317.1 | 5 | 108 | |||
| XM_011517017.2 | XP_011515319.1 | 4 | 108 | |||
| XM_017013384.1 | XP_016868873.1 | 2 | 108 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ribosomal protein L8 | RPL8 | L8 | -16.4kb | Sense | Chr. 8 - 144789765 - 144792446 on NCBI Build GRCh38 |
| Downstream | zinc finger protein 7 | ZNF7 |
HF.16, KOX4, zf30 |
16.4kb | Anti-sense | Chr. 8 - 144827500 - 144848070 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3282688 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3282683 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 20422 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 21803 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3282688 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3282683 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1700012299.hg.1 | ENST00000627453.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012299.hg.1 | ENST00000627453.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012299.hg.1 | ENST00000627453.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012299.hg.1 | ENST00000627453.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012299.hg.1 | ENST00000627453.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012299.hg.1 | ENST00000627596.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012299.hg.1 | ENST00000627596.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012299.hg.1 | ENST00000627957.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012299.hg.1 | ENST00000627957.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012299.hg.1 | ENST00000627957.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012299.hg.1 | ENST00000627957.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012299.hg.1 | ENST00000627957.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012299.hg.1 | ENST00000627957.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012299.hg.1 | ENST00000627957.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012299.hg.1 | ENST00000629311.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012299.hg.1 | ENST00000629311.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012299.hg.1 | ENST00000629311.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012299.hg.1 | ENST00000629311.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012299.hg.1 | ENST00000629311.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012299.hg.1 | ENST00000630575.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012299.hg.1 | ENST00000630575.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012299.hg.1 | ENST00000630575.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012299.hg.1 | ENST00000630575.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012299.hg.1 | ENST00000630575.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009289.hg.1 | ENST00000359971.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009289.hg.1 | ENST00000359971.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009289.hg.1 | ENST00000359971.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009289.hg.1 | ENST00000359971.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009289.hg.1 | ENST00000359971.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009289.hg.1 | ENST00000359971.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009289.hg.1 | ENST00000359971.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009289.hg.1 | ENST00000359971.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009289.hg.1 | ENST00000359971.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009289.hg.1 | ENST00000525105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009289.hg.1 | ENST00000525105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009289.hg.1 | ENST00000525105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009289.hg.1 | ENST00000525105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009289.hg.1 | ENST00000525105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009289.hg.1 | ENST00000525105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009289.hg.1 | ENST00000525105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009289.hg.1 | ENST00000525105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009289.hg.1 | ENST00000526178.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009289.hg.1 | ENST00000526178.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009289.hg.1 | ENST00000526178.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009289.hg.1 | ENST00000526178.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009289.hg.1 | ENST00000526178.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009289.hg.1 | ENST00000528012.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009289.hg.1 | ENST00000528012.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009289.hg.1 | ENST00000528012.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009289.hg.1 | ENST00000528012.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009289.hg.1 | ENST00000528012.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009289.hg.1 | ENST00000529429.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009289.hg.1 | ENST00000529429.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009289.hg.1 | ENST00000529429.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009289.hg.1 | ENST00000529429.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009289.hg.1 | ENST00000529429.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009289.hg.1 | ENST00000529429.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009289.hg.1 | ENST00000530879.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009289.hg.1 | ENST00000530879.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009289.hg.1 | ENST00000530879.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009289.hg.1 | ENST00000530879.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009289.hg.1 | ENST00000530879.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009289.hg.1 | ENST00000530879.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009289.hg.1 | ENST00000530879.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009289.hg.1 | ENST00000530879.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009289.hg.1 | ENST00000530879.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009289.hg.1 | ENST00000530879.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009289.hg.1 | ENST00000531720.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009289.hg.1 | ENST00000531720.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009289.hg.1 | ENST00000531720.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009289.hg.1 | ENST00000531720.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009289.hg.1 | ENST00000531720.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009289.hg.1 | ENST00000531720.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009289.hg.1 | ENST00000531720.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009289.hg.1 | ENST00000533965.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009289.hg.1 | ENST00000533965.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009289.hg.1 | ENST00000533965.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009289.hg.1 | ENST00000533965.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009289.hg.1 | ENST00000533965.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009289.hg.1 | ENST00000533965.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009289.hg.1 | ENST00000533965.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009289.hg.1 | ENST00000533965.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009289.hg.1 | NM_213605.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009289.hg.1 | NM_213605.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009289.hg.1 | NM_213605.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009289.hg.1 | NM_213605.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009289.hg.1 | NM_213605.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009289.hg.1 | NM_213605.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009289.hg.1 | NM_213605.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009289.hg.1 | NM_213605.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009289.hg.1 | NM_213605.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006507.hg.1 | ENST00000414438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006507.hg.1 | ENST00000414438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006507.hg.1 | ENST00000414438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006507.hg.1 | ENST00000414438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006507.hg.1 | ENST00000414603.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006507.hg.1 | ENST00000414603.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006507.hg.1 | ENST00000414438.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006507.hg.1 | ENST00000414438.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006507.hg.1 | ENST00000414438.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006507.hg.1 | ENST00000414438.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006507.hg.1 | ENST00000414603.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006507.hg.1 | ENST00000414603.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009289.hg.1 | ENST00000359971.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009289.hg.1 | ENST00000359971.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009289.hg.1 | ENST00000359971.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009289.hg.1 | ENST00000359971.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009289.hg.1 | ENST00000359971.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009289.hg.1 | ENST00000359971.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009289.hg.1 | ENST00000359971.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009289.hg.1 | ENST00000359971.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009289.hg.1 | ENST00000359971.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009289.hg.1 | ENST00000525105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009289.hg.1 | ENST00000525105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009289.hg.1 | ENST00000525105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009289.hg.1 | ENST00000525105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009289.hg.1 | ENST00000525105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009289.hg.1 | ENST00000525105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009289.hg.1 | ENST00000525105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009289.hg.1 | ENST00000525105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009289.hg.1 | ENST00000526178.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009289.hg.1 | ENST00000526178.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009289.hg.1 | ENST00000526178.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009289.hg.1 | ENST00000526178.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009289.hg.1 | ENST00000526178.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009289.hg.1 | ENST00000528012.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009289.hg.1 | ENST00000528012.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009289.hg.1 | ENST00000528012.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009289.hg.1 | ENST00000528012.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009289.hg.1 | ENST00000528012.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009289.hg.1 | ENST00000529429.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009289.hg.1 | ENST00000529429.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009289.hg.1 | ENST00000529429.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009289.hg.1 | ENST00000529429.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009289.hg.1 | ENST00000529429.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009289.hg.1 | ENST00000529429.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009289.hg.1 | ENST00000530879.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009289.hg.1 | ENST00000530879.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009289.hg.1 | ENST00000530879.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009289.hg.1 | ENST00000530879.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009289.hg.1 | ENST00000530879.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009289.hg.1 | ENST00000530879.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009289.hg.1 | ENST00000530879.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009289.hg.1 | ENST00000530879.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009289.hg.1 | ENST00000530879.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009289.hg.1 | ENST00000530879.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009289.hg.1 | ENST00000531720.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009289.hg.1 | ENST00000531720.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009289.hg.1 | ENST00000531720.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009289.hg.1 | ENST00000531720.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009289.hg.1 | ENST00000531720.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009289.hg.1 | ENST00000531720.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009289.hg.1 | ENST00000531720.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009289.hg.1 | ENST00000533965.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009289.hg.1 | ENST00000533965.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009289.hg.1 | ENST00000533965.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009289.hg.1 | ENST00000533965.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009289.hg.1 | ENST00000533965.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009289.hg.1 | ENST00000533965.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009289.hg.1 | ENST00000533965.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009289.hg.1 | ENST00000533965.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009289.hg.1 | NM_213605.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009289.hg.1 | NM_213605.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009289.hg.1 | NM_213605.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009289.hg.1 | NM_213605.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009289.hg.1 | NM_213605.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009289.hg.1 | NM_213605.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009289.hg.1 | NM_213605.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009289.hg.1 | NM_213605.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009289.hg.1 | NM_213605.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012299.hg.1 | ENST00000627453.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012299.hg.1 | ENST00000627453.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012299.hg.1 | ENST00000627453.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012299.hg.1 | ENST00000627453.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012299.hg.1 | ENST00000627453.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012299.hg.1 | ENST00000627596.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012299.hg.1 | ENST00000627596.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012299.hg.1 | ENST00000627957.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012299.hg.1 | ENST00000627957.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012299.hg.1 | ENST00000627957.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012299.hg.1 | ENST00000627957.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012299.hg.1 | ENST00000627957.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012299.hg.1 | ENST00000627957.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012299.hg.1 | ENST00000627957.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012299.hg.1 | ENST00000629311.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012299.hg.1 | ENST00000629311.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012299.hg.1 | ENST00000629311.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012299.hg.1 | ENST00000629311.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012299.hg.1 | ENST00000629311.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012299.hg.1 | ENST00000630575.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012299.hg.1 | ENST00000630575.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012299.hg.1 | ENST00000630575.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012299.hg.1 | ENST00000630575.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012299.hg.1 | ENST00000630575.2 | Ensembl |
| Human Genome U95 Set | 72655_PM_at | - | - |
| Human Genome U95 Set | 72655_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 230441 | - | - |

LOC100130027
Genomic Map