Assay Details
| Gene Symbol: LOC100132612 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK128397.1 | - | 2 | 2950 | 79 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC100129345 | LOC100129345 | -62.6kb | Sense | Chr. 14 - 97632647 - 97686658 on NCBI Build GRCh38 | |
| Downstream | long intergenic non-protein coding RNA 1550 | LINC01550 | C14orf64 | 173.6kb | Sense | Chr. 14 - 97925610 - 97978124 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09500876 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29962519 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09500862 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29962634 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09500877 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29962541 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09500827 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09500829 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29962585 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29962518 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29962532 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09500833 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09500868 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09500871 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09500854 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16731825 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29962621 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09500786 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09500885 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09500823 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29962529 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09500761 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09500811 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29962582 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09500841 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29962540 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29962537 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09500879 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09500847 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29962586 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09500760 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09500792 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29962563 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09500822 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09500839 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29962640 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09500755 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29962524 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29962569 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09500745 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09500844 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29962635 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09500778 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29962591 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29962589 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09500805 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09500830 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09500873 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29962530 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09500757 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09500771 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09500809 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09500794 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29962607 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09500803 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09500739 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09500783 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09500781 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09500810 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09500835 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29962654 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09500881 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09500818 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29962522 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09500774 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29962612 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09500859 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09500806 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09500872 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29962545 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29962628 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09500799 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09500756 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29962597 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29962542 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29962583 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09500851 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29962543 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09500751 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29962564 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09500875 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09500773 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09500815 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09500865 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29962611 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29962636 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29962555 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29962565 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29962627 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09500882 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29962598 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09500780 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09500782 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09500776 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29962544 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29962526 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29962602 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09500779 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09500801 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29962528 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09500743 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09500874 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09500824 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29962648 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09500798 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09500793 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09500860 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09500842 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09500857 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3349030 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 21890 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P02979912 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P40301063 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3349030 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P02979912 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P40301050 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P40301039 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P02979912 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P20154931 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P40301063 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P33606585 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P20154907 | - | - |

LOC100132612
Genomic Map