Assay Details
| Gene Symbol: KCNIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK130689.1 | - | 1 | 1547 | 86 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | KCNIP2 antisense RNA 1 | KCNIP2-AS1 | -8.7kb | Anti-sense | Chr. 10 - 101819068 - 101828889 on NCBI Build GRCh38 | |
| Downstream | chromosome 10 open reading frame 76 | C10orf76 | 6.3kb | Sense | Chr. 10 - 101845599 - 102056237 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

KCNIP2
Genomic Map