Assay Details
| Gene Symbol: LOC100129473 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK130906.1 | - | 2 | 948 | 85 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC26070 | DKFZP434K028 | -9.0kb | Anti-sense | Chr. 11 - 61754029 - 61757664 on NCBI Build GRCh38 | |
| Downstream | transmembrane protein 258 | TMEM258 | C11orf10 | 21.3kb | Anti-sense | Chr. 11 - 61789130 - 61792613 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100129473
Genomic Map