Assay Details
| Gene Symbol: LOC100128075 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK130927.1 | - | 1 | 448 | 100 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | tetratricopeptide repeat domain 8 | TTC8 |
BBS8, RP51 |
-389.5kb | Anti-sense | Chr. 14 - 88824153 - 88877996 on NCBI Build GRCh38 |
| Downstream | FOXN3 antisense RNA 1 | FOXN3-AS1 | 148.3kb | Anti-sense | Chr. 14 - 89417354 - 89419793 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100128075
Genomic Map