Assay Details
| Gene Symbol: DISC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AL049973.2 | - | 1 | 852 | 78 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | TSNAX-DISC1 readthrough (NMD candidate) | TSNAX-DISC1 | Sense | Chr. 1 - 231528653 - 232041272 on NCBI Build GRCh38 | ||
| Downstream | DISC1 intronic transcript 1 | DISC1-IT1 | 59.2kb | Sense | Chr. 1 - 231925834 - 231945205 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

DISC1
Genomic Map