Assay Details
| Gene Symbol: LOC286191 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AL833212.1 | - | 1 | 844 | 89 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | telomeric repeat binding factor 1 | TERF1 |
PIN2, TRBF1, TRF, TRF1, hTRF1-AS, t-TRF1 |
-13.3kb | Anti-sense | Chr. 8 - 73008862 - 73047752 on NCBI Build GRCh38 |
| Downstream | somatomedin B and thrombospondin type 1 domain containing | SBSPON |
C8orf84, RPESP |
1.3kb | Sense | Chr. 8 - 73063077 - 73093272 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC286191
Genomic Map