Assay Details
| Gene Symbol: FARP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC036887.1 | - | 2 | 1951 | 90 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 3133 | MIR3133 | -4.0kb | Anti-sense | Chr. 2 - 241477905 - 241477982 on NCBI Build GRCh38 | |
| Downstream | serine/threonine kinase 25 | STK25 |
SOK1, YSK1 |
11.7kb | Sense | Chr. 2 - 241494707 - 241509730 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

FARP2
Genomic Map