Assay Details
| Gene Symbol: ZDHHC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC045169.2 | - | 2 | 2153 | 98 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | tubulin polymerization promoting protein family member 3 | TPPP3 |
CGI-38, TPPP/p20, p20, p25gamma |
-5.5kb | Anti-sense | Chr. 16 - 67389807 - 67393535 on NCBI Build GRCh38 |
| Downstream | hydroxysteroid 11-beta dehydrogenase 2 | HSD11B2 |
AME, AME1, HSD11K, HSD2, SDR9C3 |
30.6kb | Sense | Chr. 16 - 67431133 - 67437553 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

ZDHHC1
Genomic Map