Assay Details
| Gene Symbol: UBN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC045656.1 | - | 1 | 3147 | 79 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | tetratricopeptide repeat domain 26 | TTC26 |
DYF13, IFT56, dyf-13 |
-63.9kb | Anti-sense | Chr. 7 - 139133744 - 139191986 on NCBI Build GRCh38 |
| Downstream |
LUC7 like 2, pre-mRNA splicing factor |
LUC7L2 |
CGI-59, CGI-74, LUC7B2 |
81.0kb | Anti-sense | Chr. 7 - 139340359 - 139423457 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

UBN2
Genomic Map