Assay Details
| Gene Symbol: PSPH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK054677.1 | - | 5 | 1721 | 114 | |
| BC052958.1 | - | 3 | 2651 | 114 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | glioblastoma amplified sequence | GBAS | NIPSNAP2 | -40.2kb | Anti-sense | Chr. 7 - 55964577 - 56000182 on NCBI Build GRCh38 |
| Downstream | chaperonin containing TCP1 subunit 6A | CCT6A |
CCT-zeta, CCT-zeta-1, CCT6, Cctz, HTR3, MoDP-2, TCP-1-zeta, TCP20, TCPZ, TTCP20 |
9.0kb | Anti-sense | Chr. 7 - 56051685 - 56063989 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

PSPH
Genomic Map