Assay Details
| Gene Symbol: LDLR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | M28219.1 | - | 1 | 46 | 80 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 6886 | MIR6886 | hsa-mir-6886 | -9.8kb | Sense | Chr. 19 - 11113474 - 11113534 on NCBI Build GRCh38 |
| Downstream |
SPC24, NDC80 kinetochore complex component |
SPC24 | SPBC24 | 21.9kb | Anti-sense | Chr. 19 - 11145494 - 11155997 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LDLR
Genomic Map