Assay Details
| Gene Symbol: CACNB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK022426.1 | - | 1 | 1350 | 73 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | solute carrier family 39 member 12 | SLC39A12 |
LZT-Hs8, ZIP-12, bA570F3.1 |
-253.3kb | Sense | Chr. 10 - 17951839 - 18043292 on NCBI Build GRCh38 |
| Downstream | NOP2/Sun RNA methyltransferase family member 6 | NSUN6 |
4933414E04Rik, ARL5B-AS1, NOPD1 |
246.8kb | Anti-sense | Chr. 10 - 18512582 - 18659264 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

CACNB2
Genomic Map