Assay Details
| Gene Symbol: ORMDL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK126336.1 | - | 1 | 2043 | 113 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | OSGEPL1 antisense RNA 1 | OSGEPL1-AS1 | -5.8kb | Anti-sense | Chr. 2 - 189762780 - 189765556 on NCBI Build GRCh38 | |
| Downstream |
PMS1 homolog 1, mismatch repair system component |
PMS1 |
HNPCC3, MLH2, PMSL1, hPMS1 |
8.7kb | Anti-sense | Chr. 2 - 189784085 - 189877629 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

ORMDL1
Genomic Map