Assay Details
| Gene Symbol: ZBTB16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK126422.1 | - | 1 | 1751 | 76 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | 5-hydroxytryptamine receptor 3A | HTR3A |
5-HT-3, 5-HT3A, 5-HT3R, 5HT3R, HTR3 |
-104.2kb | Sense | Chr. 11 - 113975075 - 113990313 on NCBI Build GRCh38 |
| Downstream | nicotinamide N-methyltransferase | NNMT | 199.2kb | Sense | Chr. 11 - 114295813 - 114312516 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

ZBTB16
Genomic Map