Assay Details
| Gene Symbol: CYP39A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC012548.1 | - | 1 | 1751 | 113 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101926898 | LOC101926898 | -117.0kb | Anti-sense | Chr. 6 - 46492052 - 46533486 on NCBI Build GRCh38 | |
| Downstream | solute carrier family 25 member 27 | SLC25A27 | UCP4 | 165.0 | Anti-sense | Chr. 6 - 46652915 - 46678190 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

CYP39A1
Genomic Map