Assay Details
| Gene Symbol: STXBP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC028028.1 | - | 1 | 744 | 71 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | fibronectin type III domain containing 7 | FNDC7 | -3.4kb | Sense | Chr. 1 - 108712934 - 108742749 on NCBI Build GRCh38 | |
| Downstream | AKNA domain containing 1 | AKNAD1 | C1orf62 | 67.7kb | Anti-sense | Chr. 1 - 108815898 - 108858242 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

STXBP3
Genomic Map