Assay Details
| Gene Symbol: LINC01126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027251.1 | 1 | 181 | 110 | ||
| GenBank mRNA | BC032412.1 | - | 1 | 51 | 5216225 | 110 |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ZFP36 ring finger protein like 2 | ZFP36L2 |
BRF2, ERF-2, ERF2, RNF162C, TIS11D |
-735.0 | Anti-sense | Chr. 2 - 43222402 - 43226606 on NCBI Build GRCh38 |
| Downstream |
THADA, armadillo repeat containing |
THADA |
ARMC13, GITA |
2.0kb | Anti-sense | Chr. 2 - 43230836 - 43596046 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02000267.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC032412.1 transcripts.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 123660 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 8x60K | 51540 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 696152 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P348911 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P348911 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_5074 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_5073 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 28069 | - | - |
| Agilent Human Genome CGH Microarray 44B | A_14_P117889 | - | - |
| Agilent Human Genome CGH Microarray 44A | A_14_P117889 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 9509 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P348911 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_23_P348911 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21216232 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01135964 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01135969 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01135965 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21216222 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01135980 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01135961 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01135963 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01135962 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01135967 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01135975 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01135971 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01135968 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01135977 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21216226 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21216231 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01135962 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01135977 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21216232 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01135967 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21216231 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01135975 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21216226 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01135971 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01135968 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21216222 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01135961 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01135969 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01135963 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01135965 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01135980 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01135964 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_23_P348911 | - | - |
| Agilent Human Genome CGH Microarray 105A (G4412A) | A_14_P117889 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_14_P117889 | - | - |
| Agilent Human Genome CGH Microarray 4x44K | A_14_P117889 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P348911 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 164316 | - | - |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 8110 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_14_P117889 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01135964 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01135962 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01135967 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01135979 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01135969 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01135976 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01135980 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01135973 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01135971 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15237734 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01135961 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01135963 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01135965 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_14_P117889 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 8041641 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8041640 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 1555124_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11749570_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11749570_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003960.hg.1 | TC02000267.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003960.hg.1 | NR_027251.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003961.hg.1 | TC02000267.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003961.hg.1 | NR_027251.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007388.hg.1 | ENST00000623515.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007388.hg.1 | ENST00000623515.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007388.hg.1 | NR_027251.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007388.hg.1 | NR_027251.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200151143.hg.1 | ENST00000623515.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200151143.hg.1 | NR_027251.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200151143.hg.1 | ENST00000623515.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200151143.hg.1 | NR_027251.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200151143.hg.1 | TC0200007388.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200007388.hg.1 | ENST00000623515.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007388.hg.1 | ENST00000623515.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007388.hg.1 | NR_027251.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007388.hg.1 | NR_027251.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200151142.hg.1 | ENST00000623515.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200151142.hg.1 | NR_027251.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200151142.hg.1 | ENST00000623515.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200151142.hg.1 | NR_027251.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200151142.hg.1 | TC0200007388.hg.1 | Clariom D TC |
| Human Genome U133 Plus 2.0 Array | 1555124_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1555124_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1555124_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1555124_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1555124_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1555124_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1555124_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1555124_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1555124_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1555124_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1555124_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1555124_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1555124_at3 | - | - |
| Human Genome U95 Set | 86219_PM_at | - | - |
| Human Genome U95 Set | 86219_at | - | - |
| Human Genome U133 Set | 231189_PM_at | - | - |
| Human Genome U133 Set | 231189_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A015577 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A015577 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 221725 | - | - |
| ABI Human Genome Survey Microarray Version 2.0 | 235098 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3187435 | NR_027251.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3187435 | BC032412.1 | human_transcriptome |

LINC01126
Genomic Map