Assay Details
| Gene Symbol: PFKFB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC042656.1 | - | 1 | 555 | 93 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 3155a | MIR3155A |
MIR3155, hsa-mir-3155a, mir-3155a |
-78.7kb | Sense | Chr. 10 - 6152196 - 6152277 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC399715 | LOC399715 | 91.0kb | Sense | Chr. 10 - 6326544 - 6335982 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

PFKFB3
Genomic Map