Assay Details
| Gene Symbol: EDEM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC070173.1 | - | 1 | 750 | 100 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | chromosome 1 open reading frame 21 | C1orf21 | PIG13 | -124.2kb | Anti-sense | Chr. 1 - 184387016 - 184629021 on NCBI Build GRCh38 |
| Downstream | family with sequence similarity 129 member A | FAM129A |
C1orf24, GIG39, NIBAN |
36.2kb | Sense | Chr. 1 - 184791025 - 184975132 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

EDEM3
Genomic Map