Assay Details
| Gene Symbol: DIRC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_026597.1 | 2 - 3 | 1349 | 93 | ||
| GenBank mRNA | AK024261.1 | - | 1 - 2 | 326 | 93 | |
| AK090954.1 | - | 2 - 3 | 1352 | 93 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | DIRC3 antisense RNA 1 | DIRC3-AS1 | -158.8kb | Anti-sense | Chr. 2 - 217282733 - 217344204 on NCBI Build GRCh38 | |
| Downstream | tensin 1 | TNS1 |
MST091, MST122, MST127, MSTP091, MSTP122, MSTP127, MXRA6, PPP1R155, TNS |
199.0kb | Sense | Chr. 2 - 217799789 - 217978853 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02002764.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02044364.hg.1 | TC02002764.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02044364.hg.1 | NR_026597.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015695.hg.1 | ENST00000423123.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015695.hg.1 | ENST00000423123.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015695.hg.1 | ENST00000423123.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015695.hg.1 | ENST00000423123.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015695.hg.1 | ENST00000423123.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015695.hg.1 | ENST00000474063.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015695.hg.1 | ENST00000474063.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015695.hg.1 | ENST00000474063.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015695.hg.1 | ENST00000474063.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015695.hg.1 | ENST00000474063.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015695.hg.1 | ENST00000474063.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015695.hg.1 | ENST00000474063.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015695.hg.1 | ENST00000474063.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015695.hg.1 | ENST00000474063.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015695.hg.1 | ENST00000474063.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015695.hg.1 | ENST00000474063.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015695.hg.1 | ENST00000474063.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015695.hg.1 | ENST00000474063.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015695.hg.1 | ENST00000484635.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015695.hg.1 | ENST00000484635.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015695.hg.1 | ENST00000484635.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015695.hg.1 | ENST00000484635.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015695.hg.1 | ENST00000484635.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015695.hg.1 | ENST00000486365.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015695.hg.1 | ENST00000486365.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015695.hg.1 | ENST00000486365.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015695.hg.1 | ENST00000486365.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015695.hg.1 | ENST00000486365.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015695.hg.1 | ENST00000486365.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015695.hg.1 | ENST00000486365.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015695.hg.1 | ENST00000486365.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015695.hg.1 | ENST00000486365.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015695.hg.1 | ENST00000486365.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015695.hg.1 | ENST00000486365.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015695.hg.1 | ENST00000486365.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015695.hg.1 | NR_026597.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015695.hg.1 | NR_026597.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015695.hg.1 | NR_026597.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015695.hg.1 | NR_026597.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015695.hg.1 | NR_026597.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015695.hg.1 | NR_026597.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015695.hg.1 | NR_026597.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015695.hg.1 | NR_026597.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015695.hg.1 | NR_026597.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015695.hg.1 | NR_026597.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015695.hg.1 | NR_026597.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015695.hg.1 | NR_026597.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015695.hg.1 | NR_026597.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015695.hg.1 | NR_026597.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015695.hg.1 | NR_026597.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02044363.hg.1 | TC02002764.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02044363.hg.1 | ENST00000423123.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02044363.hg.1 | NR_026597.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02044357.hg.1 | TC02002764.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02044357.hg.1 | ENST00000474063.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02044357.hg.1 | NR_026597.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02044356.hg.1 | TC02002764.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02044356.hg.1 | ENST00000474063.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02044356.hg.1 | NR_026597.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02044358.hg.1 | TC02002764.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02044358.hg.1 | NR_026597.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015695.hg.1 | ENST00000423123.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015695.hg.1 | ENST00000423123.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015695.hg.1 | ENST00000423123.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015695.hg.1 | ENST00000423123.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015695.hg.1 | ENST00000423123.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015695.hg.1 | ENST00000474063.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015695.hg.1 | ENST00000474063.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015695.hg.1 | ENST00000474063.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015695.hg.1 | ENST00000474063.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015695.hg.1 | ENST00000474063.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015695.hg.1 | ENST00000474063.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015695.hg.1 | ENST00000474063.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015695.hg.1 | ENST00000474063.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015695.hg.1 | ENST00000474063.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015695.hg.1 | ENST00000474063.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015695.hg.1 | ENST00000474063.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015695.hg.1 | ENST00000474063.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015695.hg.1 | ENST00000474063.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015695.hg.1 | ENST00000484635.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015695.hg.1 | ENST00000484635.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015695.hg.1 | ENST00000484635.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015695.hg.1 | ENST00000484635.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015695.hg.1 | ENST00000484635.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015695.hg.1 | ENST00000486365.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015695.hg.1 | ENST00000486365.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015695.hg.1 | ENST00000486365.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015695.hg.1 | ENST00000486365.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015695.hg.1 | ENST00000486365.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015695.hg.1 | ENST00000486365.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015695.hg.1 | ENST00000486365.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015695.hg.1 | ENST00000486365.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015695.hg.1 | ENST00000486365.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015695.hg.1 | ENST00000486365.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015695.hg.1 | ENST00000486365.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015695.hg.1 | ENST00000486365.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015695.hg.1 | NR_026597.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015695.hg.1 | NR_026597.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015695.hg.1 | NR_026597.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015695.hg.1 | NR_026597.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015695.hg.1 | NR_026597.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015695.hg.1 | NR_026597.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015695.hg.1 | NR_026597.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015695.hg.1 | NR_026597.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015695.hg.1 | NR_026597.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015695.hg.1 | NR_026597.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015695.hg.1 | NR_026597.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015695.hg.1 | NR_026597.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015695.hg.1 | NR_026597.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015695.hg.1 | NR_026597.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015695.hg.1 | NR_026597.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200215182.hg.1 | ENST00000486365.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200215182.hg.1 | NR_026597.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200215182.hg.1 | ENST00000486365.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200215182.hg.1 | NR_026597.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200215182.hg.1 | TC0200015695.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200215187.hg.1 | ENST00000423123.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200215187.hg.1 | ENST00000486365.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200215187.hg.1 | NR_026597.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200215187.hg.1 | ENST00000423123.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200215187.hg.1 | ENST00000486365.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200215187.hg.1 | NR_026597.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200215187.hg.1 | TC0200015695.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200215181.hg.1 | ENST00000474063.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200215181.hg.1 | ENST00000486365.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200215181.hg.1 | NR_026597.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200215181.hg.1 | ENST00000474063.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200215181.hg.1 | ENST00000486365.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200215181.hg.1 | NR_026597.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200215181.hg.1 | TC0200015695.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200215188.hg.1 | ENST00000486365.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200215188.hg.1 | NR_026597.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200215188.hg.1 | ENST00000486365.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200215188.hg.1 | NR_026597.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200215188.hg.1 | TC0200015695.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200215180.hg.1 | ENST00000474063.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200215180.hg.1 | ENST00000486365.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200215180.hg.1 | NR_026597.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200215180.hg.1 | ENST00000474063.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200215180.hg.1 | ENST00000486365.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200215180.hg.1 | NR_026597.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200215180.hg.1 | TC0200015695.hg.1 | Clariom D TC |

DIRC3
Genomic Map