| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02044351.hg.1 |
TC02002764.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02044351.hg.1 |
ENST00000474063.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02044351.hg.1 |
NR_026597.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015695.hg.1 |
ENST00000423123.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015695.hg.1 |
ENST00000423123.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015695.hg.1 |
ENST00000423123.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015695.hg.1 |
ENST00000423123.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015695.hg.1 |
ENST00000423123.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015695.hg.1 |
ENST00000474063.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015695.hg.1 |
ENST00000474063.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015695.hg.1 |
ENST00000474063.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015695.hg.1 |
ENST00000474063.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015695.hg.1 |
ENST00000474063.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015695.hg.1 |
ENST00000474063.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015695.hg.1 |
ENST00000474063.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015695.hg.1 |
ENST00000474063.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015695.hg.1 |
ENST00000474063.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015695.hg.1 |
ENST00000474063.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015695.hg.1 |
ENST00000474063.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015695.hg.1 |
ENST00000474063.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015695.hg.1 |
ENST00000474063.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015695.hg.1 |
ENST00000484635.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015695.hg.1 |
ENST00000484635.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015695.hg.1 |
ENST00000484635.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015695.hg.1 |
ENST00000484635.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015695.hg.1 |
ENST00000484635.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015695.hg.1 |
ENST00000486365.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015695.hg.1 |
ENST00000486365.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015695.hg.1 |
ENST00000486365.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015695.hg.1 |
ENST00000486365.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015695.hg.1 |
ENST00000486365.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015695.hg.1 |
ENST00000486365.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015695.hg.1 |
ENST00000486365.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015695.hg.1 |
ENST00000486365.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015695.hg.1 |
ENST00000486365.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015695.hg.1 |
ENST00000486365.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015695.hg.1 |
ENST00000486365.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015695.hg.1 |
ENST00000486365.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015695.hg.1 |
NR_026597.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015695.hg.1 |
NR_026597.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015695.hg.1 |
NR_026597.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015695.hg.1 |
NR_026597.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015695.hg.1 |
NR_026597.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015695.hg.1 |
NR_026597.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015695.hg.1 |
NR_026597.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015695.hg.1 |
NR_026597.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015695.hg.1 |
NR_026597.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015695.hg.1 |
NR_026597.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015695.hg.1 |
NR_026597.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015695.hg.1 |
NR_026597.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015695.hg.1 |
NR_026597.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015695.hg.1 |
NR_026597.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015695.hg.1 |
NR_026597.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015695.hg.1 |
ENST00000423123.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015695.hg.1 |
ENST00000423123.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015695.hg.1 |
ENST00000423123.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015695.hg.1 |
ENST00000423123.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015695.hg.1 |
ENST00000423123.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015695.hg.1 |
ENST00000474063.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015695.hg.1 |
ENST00000474063.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015695.hg.1 |
ENST00000474063.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015695.hg.1 |
ENST00000474063.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015695.hg.1 |
ENST00000474063.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015695.hg.1 |
ENST00000474063.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015695.hg.1 |
ENST00000474063.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015695.hg.1 |
ENST00000474063.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015695.hg.1 |
ENST00000474063.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015695.hg.1 |
ENST00000474063.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015695.hg.1 |
ENST00000474063.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015695.hg.1 |
ENST00000474063.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015695.hg.1 |
ENST00000474063.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015695.hg.1 |
ENST00000484635.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015695.hg.1 |
ENST00000484635.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015695.hg.1 |
ENST00000484635.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015695.hg.1 |
ENST00000484635.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015695.hg.1 |
ENST00000484635.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015695.hg.1 |
ENST00000486365.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015695.hg.1 |
ENST00000486365.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015695.hg.1 |
ENST00000486365.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015695.hg.1 |
ENST00000486365.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015695.hg.1 |
ENST00000486365.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015695.hg.1 |
ENST00000486365.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015695.hg.1 |
ENST00000486365.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015695.hg.1 |
ENST00000486365.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015695.hg.1 |
ENST00000486365.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015695.hg.1 |
ENST00000486365.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015695.hg.1 |
ENST00000486365.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015695.hg.1 |
ENST00000486365.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015695.hg.1 |
NR_026597.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015695.hg.1 |
NR_026597.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015695.hg.1 |
NR_026597.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015695.hg.1 |
NR_026597.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015695.hg.1 |
NR_026597.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015695.hg.1 |
NR_026597.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015695.hg.1 |
NR_026597.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015695.hg.1 |
NR_026597.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015695.hg.1 |
NR_026597.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015695.hg.1 |
NR_026597.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015695.hg.1 |
NR_026597.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015695.hg.1 |
NR_026597.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015695.hg.1 |
NR_026597.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015695.hg.1 |
NR_026597.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015695.hg.1 |
NR_026597.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200215174.hg.1 |
ENST00000474063.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200215174.hg.1 |
ENST00000484635.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200215174.hg.1 |
ENST00000486365.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200215174.hg.1 |
NR_026597.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200215174.hg.1 |
ENST00000474063.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200215174.hg.1 |
ENST00000484635.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200215174.hg.1 |
ENST00000486365.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200215174.hg.1 |
NR_026597.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200215174.hg.1 |
TC0200015695.hg.1 |
Clariom D TC |